tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals
CAS: 1948-33-0 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
Packaging:
View all
Plastic bottle
Catalog number ACR150822500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS497-19-8
IUPAC Namedisodium carbonate
Molecular FormulaCNa2O3
InChI KeyCDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<10 ppm
Appearance (Color)White
Appearance (Form)Crystalline powder