2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,3-Butanedione monoxime, 98%, Thermo Scientific Chemicals
CAS: 57-71-6 | C4H7NO2 | 101.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number ACR150371000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 3-Butanedione monoxime
CAS57-71-6
Melting Point74°C to 76°C
ColorWhite
EINECS Number200-348-5