Ethyl acetate, 99+%, Thermo Scientific Chemicals
Ethyl acetate, 99+%, Thermo Scientific Chemicals
Ethyl acetate, 99+%, Thermo Scientific Chemicals
Ethyl acetate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl acetate, 99+%, Thermo Scientific Chemicals
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number ACR149470010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS9048-46-8
MDL NumberMFCD00145743
SpecificationsSpecification SheetSpecification Sheet
Sulfated Ash5.0% max.
Assay (unspecified)Moisture: 5.0% max.
Assay (unspecified)Protein: 97% min.
pH4.5-5.5, 10% solution