Phenol, 99%, extra pure, Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenol, 99%, extra pure, Thermo Scientific Chemicals
CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
50 g
1 kg
Packaging:
View all
Plastic drum
Catalog number ACR149340051
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS1312-76-1
IUPAC Namepotassium hydroxysilanoylolate
Molecular FormulaHKO3Si
InChI KeyWMWBFVSTSRDWRL-UHFFFAOYSA-N
SMILES[K+].O[Si]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)28.0-28.8% K₂O