Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
CAS: 367-51-1 | C2H3NaO2S | 114.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number ACR148890500
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMercaptoacetic acid, sodium salt
Name Note98%
CAS367-51-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be corrosive to metals.