Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mercaptoacetic acid, sodium salt, 98%, Thermo Scientific Chemicals
CAS: 367-51-1 | C2H3NaO2S | 114.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Packaging:
Glass bottle
Catalog number ACR148890010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3069-40-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)> 97.0%