Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number ACR146750010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthylene glycol
Melting Point-13°C
Name Note99+%
CAS107-21-1
Health Hazard 1Warning