1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
Thermo Scientific Chemicals
1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number ACR146310050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-(2-Pyridylazo)-2-naphthol
CAS85-85-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
View more
Indicator
RUO – Research Use Only