delta-Valerolactam, 99%, Thermo Scientific Chemicals
delta-Valerolactam, 99%, Thermo Scientific Chemicals
delta-Valerolactam, 99%, Thermo Scientific Chemicals
delta-Valerolactam, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
delta-Valerolactam, 99%, Thermo Scientific Chemicals
CAS: 675-20-7 | C5H9NO | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR140780050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS481681-02-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%