1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals
1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals
1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals
1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,4-1H-Triazole, 99.5%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
View all
Glass Bottle
Catalog number ACR139280050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 2, 4-1H-Triazole
CAS288-88-0
CAS Min %99.4
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0