p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Toluenesulfonyl chloride, 99+%, Thermo Scientific Chemicals
CAS: 98-59-9 | C7H7ClO2S | 190.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR139030020
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS75-77-4
IUPAC Namechlorotrimethylsilane
Molecular FormulaC3H9ClSi
InChI KeyIJOOHPMOJXWVHK-UHFFFAOYSA-N
SMILESC[Si](C)(C)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
Refractive Index1.3865-1.3905 @ 20°C