Check Order Status
2-Amino-6-chlorobenzothiazole, 99%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
2-Amino-6-chlorobenzothiazole, 99%, Thermo Scientific™
Have Questions?
Change view
Quantity:
10 g
Packaging:
Glass bottle
Catalog number
ACR136400100
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
2-Amino-6-chlorobenzothiazole
CAS
95-24-9
CAS Min %
98.5
Health Hazard 1
GHS Signal Word:
Warning
CAS Max %
100.0