2-chloroquinoxaline, 97%, Thermo Scientific Chemicals
2-chloroquinoxaline, 97%, Thermo Scientific Chemicals
2-chloroquinoxaline, 97%, Thermo Scientific Chemicals
2-chloroquinoxaline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-chloroquinoxaline, 97%, Thermo Scientific Chemicals
CAS: 1448-87-9 | C8H5ClN2 | 164.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR136190050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-chloroquinoxaline
CAS1448-87-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0