Activated charcoal, NORIT(R) A SUPRA, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) A SUPRA, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) A SUPRA, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) A SUPRA, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Activated charcoal, NORIT(R) A SUPRA, Thermo Scientific Chemicals
Activated charcoal,NORIT™ A SUPRA, C, CAS Number-7440-44-0, acticarbone, activated charcoal, anthrasorb, carbon-12, carbono, carbosieve, graphene, graphite, mineral, norit, 1kg, Passes Test, CHEBI:27594, Black, 231-153-3, OKTJSMMVPCPJKN-UHFFFAOYSA-N, 15% max., 3652.0 deg.C, 15, 1807
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
View all
Glass bottle
Catalog number ACR134342500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS93-05-0
IUPAC NameN1,N1-diethylbenzene-1,4-diamine
Molecular FormulaC10H16N2
InChI KeyQNGVNLMMEQUVQK-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear yellow-brown to dark brown to black
Infrared spectrumConforms
Appearance (Form)Liquid or low melting solid
Filter aid
RUO – Research Use Only