Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, H3N, CAS Number-7664-41-7, 67-56-1, am-fol, ammonia, ammonia anhydrous, ammonia, anhydrous, ammonia gas, ammoniak, ammoniakgas, anhydrous ammonia, nitro-sil, spirit of hartshorn, 2.5L, CHEBI:16134, Colorless to Yellow, 6.0 to 7.5N, 0.7700g/mL, 05,15, 14 deg.C, 17.03, GHS Signal Word: Danger
Have Questions?
Catalog NumberQuantityPackaging
ACR13371025025 mLGlass Bottle
Catalog number ACR133710250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS497-25-6
IUPAC Name1,3-oxazolidin-2-one
Molecular FormulaC3H5NO2
InChI KeyIZXIZTKNFFYFOF-UHFFFAOYSA-N
SMILESO=C1NCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥98.5%