Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, H3N, CAS Number-7664-41-7, 67-56-1, am-fol, ammonia, ammonia anhydrous, ammonia, anhydrous, ammonia gas, ammoniak, ammoniakgas, anhydrous ammonia, nitro-sil, spirit of hartshorn, 2.5L, CHEBI:16134, Colorless to Yellow, 6.0 to 7.5N, 0.7700g/mL, 05,15, 14 deg.C, 17.03, GHS Signal Word: Danger
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 mL
2.5 L
Packaging:
Glass Bottle
Catalog number ACR133710010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS93-25-4
IUPAC Name2-(2-methoxyphenyl)acetic acid
Molecular FormulaC9H10O3
InChI KeyIVEWTCACRDEAOB-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point122°C to 125°C
Appearance (Color)White to cream to light pink
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with NaOH>=98.5 %