3-Aminoisoxazole, 95%, Thermo Scientific Chemicals
3-Aminoisoxazole, 95%, Thermo Scientific Chemicals
3-Aminoisoxazole, 95%, Thermo Scientific Chemicals
3-Aminoisoxazole, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Aminoisoxazole, 95%, Thermo Scientific Chemicals
CAS: 1750-42-1 | C3H4N2O | 84.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
Packaging:
Glass bottle
Catalog number ACR133530050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS34824-58-3
IUPAC Name2-(2-bromophenyl)-1,3-dioxolane
Molecular FormulaC9H9BrO2
InChI KeyIWSGKSUCFVOWQU-UHFFFAOYSA-N
SMILESBrC1=CC=CC=C1C1OCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5670-1.5710 @ 20?C