DL-Phenylsuccinic acid, 98+%, Thermo Scientific Chemicals
DL-Phenylsuccinic acid, 98+%, Thermo Scientific Chemicals
DL-Phenylsuccinic acid, 98+%, Thermo Scientific Chemicals
DL-Phenylsuccinic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Phenylsuccinic acid, 98+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR130941000100 gGlass bottle
Catalog number ACR130941000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDL-Phenylsuccinic acid
CAS635-51-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.