2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Nitrobenzaldehyde, 99+%, Thermo Scientific Chemicals
CAS: 552-89-6 | C7H5NO3 | 151.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Catalog number ACR128401000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS75-47-8
IUPAC Nametriiodomethane
Molecular FormulaCHI3
InChI KeyOKJPEAGHQZHRQV-UHFFFAOYSA-N
SMILESIC(I)I
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Bright yellow to dark yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point119°C to 122°C
Loss on drying=<0.5 % (24 h) (Over silicagel)