4-Nitroaniline, 99%, Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
CAS: 100-01-6 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number ACR128375000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12045-78-2
IUPAC Namepotassium tetrahydrate borate
Molecular FormulaBH8KO7
InChI KeySAAWPUYPXWYTFF-UHFFFAOYSA-N
SMILESO.O.O.O.[K+].[O-]B([O-])[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<20 ppm
Appearance (Form)Powder
Titration with HClO4>=99.4 %
Heavy metals (as Pb)=<20 ppm
Chloride (Cl)=<20 ppm
View more
Indicator
RUO – Research Use Only