4-Nitroaniline, 99%, Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitroaniline, 99%, Thermo Scientific Chemicals
CAS: 100-01-6 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
View all
Plastic Bottle
Catalog number ACR128370025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS142906-29-4
IUPAC Namediiron(3+) pentahydrate trisulfate
Molecular FormulaFe2H10O17S3
InChI KeyYHGPYBQVSJBGHH-UHFFFAOYSA-H
SMILESO.O.O.O.O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline or granular powder
Insoluble matter=<0.01 % (in HCl)
Zinc (Zn)=<50 ppm
Subst not pptd by NH3=<0.05 % (as SO4)
Copper (Cu)=<20 ppm
View more
Indicator
RUO – Research Use Only