Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 75-18-3 | C2H6S | 62.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
10 mL
2.5 L
Packaging:
Glass bottle
Catalog number ACR127760010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS394-41-2
IUPAC Name3-fluoro-4-nitrobenzen-1-olate
Molecular FormulaC6H3FNO3
InChI KeyCSSGKHVRDGATJL-UHFFFAOYSA-M
SMILES[O-]C1=CC=C(C(F)=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to brown
FormPowder
Assay (GC)≥98.5%
Water Content (Karl Fischer Titration)≤0.75%
Melting Point (clear melt)86.0-95.0°C