4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
CAS: 150-76-5 | C7H8O2 | 124.14 g/mol
Have Questions?
Catalog NumberQuantity
ACR126005000500 g
Catalog number ACR126005000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS657-27-2
IUPAC Name(2S)-2,6-diaminohexanoic acid hydrochloride
Molecular FormulaC6H15ClN2O2
InChI KeyBVHLGVCQOALMSV-JEDNCBNOSA-N
SMILESCl.NCCCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Loss on drying=<0.4 % (105°C, 3 h)
Sulfate (SO4)=<300 ppm
Appearance (Color)White
Titration with HClO498.5 to 101.0 % (on dried substance)