Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Mercaptoacetic Acid, 98%, Thermo Scientific Chemicals
CAS: 68-11-1 | C2H4O2S | 92.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
10 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR125432500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6074-84-6
IUPAC Namepentaethoxytantalum
Molecular FormulaC10H25O5Ta
InChI KeyHSXKFDGTKKAEHL-UHFFFAOYSA-N
SMILESCCO[Ta](OCC)(OCC)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid (as melt)
Elemental Analysis (typical)Magnesium, Mg <20 ppm
Elemental Analysis (typical)Sodium, Na <20 ppm
Appearance (Color)Clear colorless to pale yellow
Assay (special)≥99.0% (based on tantalum EA)