DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Malic acid, +99%, Thermo Scientific Chemicals
CAS: 6915-15-7 | C4H6O5 | 134.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number ACR125252500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS39263-32-6
IUPAC Name2-amino-5-bromobenzonitrile
Molecular FormulaC7H5BrN2
InChI KeyOATYCBHROMXWJO-UHFFFAOYSA-N
SMILESNC1=CC=C(Br)C=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>95.0%