L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(+)-Lactic acid, 20 wt.% solution in water, Thermo Scientific Chemicals
CAS: 79-33-4 | C3H6O3 | 90.078 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1250700101 LGlass bottle
Catalog number ACR125070010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialL(+)-Lactic acid
CAS79-33-4
Health Hazard 1Danger
ColorColorless
Health Hazard 2GHS P Statement