Isonicotinic acid hydrazide, 99%, Thermo Scientific Chemicals
Isonicotinic acid hydrazide, 99%, Thermo Scientific Chemicals
Isonicotinic acid hydrazide, 99%, Thermo Scientific Chemicals
Isonicotinic acid hydrazide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isonicotinic acid hydrazide, 99%, Thermo Scientific Chemicals
CAS: 54-85-3 | C6H7N3O | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR122605000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS14024-58-9
IUPAC Namemanganese(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14MnO4
InChI KeyZQZQURFYFJBOCE-FDGPNNRMSA-L
SMILES[Mn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown to brown
Assay (unspecified)≥94.0 to ≤106.0% by Mn EA
Elemental AnalysisManganese Mn: 20.40 - 23.00% (Theory 21.70%)