Isobutyronitrile, 99%
Isobutyronitrile, 99%
Isobutyronitrile, 99%
Isobutyronitrile, 99%
Thermo Scientific Chemicals
Isobutyronitrile, 99%
CAS: 78-82-0 | C4H7N | 69.11 g/mol
Have Questions?
Catalog number ACR122530010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS2257-09-2
IUPAC Name(2-isothiocyanatoethyl)benzene
Molecular FormulaC9H9NS
InChI KeyIZJDOKYDEWTZSO-UHFFFAOYSA-N
SMILESS=C=NCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5860 to 1.5910 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=95 %