Isobutyric Acid, 99+%, Thermo Scientific Chemicals
Isobutyric Acid, 99+%, Thermo Scientific Chemicals
Isobutyric Acid, 99+%, Thermo Scientific Chemicals
Isobutyric Acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isobutyric Acid, 99+%, Thermo Scientific Chemicals
CAS: 79-31-2 | C4H8O2 | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number ACR122520250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIsobutyric acid
Name Note99+%
CAS79-31-2
Melting Point-47.0°C
Boiling Point153.0°C to 154.0°C
View more
Acylation reagent
RUO – Research Use Only