Isobutyric Acid, 99+%, Thermo Scientific Chemicals
Isobutyric Acid, 99+%, Thermo Scientific Chemicals
Isobutyric Acid, 99+%, Thermo Scientific Chemicals
Isobutyric Acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isobutyric Acid, 99+%, Thermo Scientific Chemicals
CAS: 79-31-2 | C4H8O2 | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Catalog number ACR122520010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS5785-44-4
IUPAC Nametricalcium bis(2-hydroxypropane-1,2,3-tricarboxylate) tetrahydrate
Molecular FormulaC12H18Ca3O18
InChI KeyLNIZKKFWMDARJV-UHFFFAOYSA-H
SMILESO.O.O.O.[Ca++].[Ca++].[Ca++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Fluoride (F)=<0.003 %
Heavy metals (as Pb)=<0.002 %
Appearance (Form)Powder
View more
Acylation reagent
RUO – Research Use Only