Formamidine acetate, 99%, Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formamidine acetate, 99%, Thermo Scientific Chemicals
CAS: 3473-63-0 | CH4N2·C2H4O2 | 104.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR119715000500 gPlastic bottle
Catalog number ACR119715000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS21615-34-9
IUPAC Name2-methoxybenzoyl chloride
Molecular FormulaC8H7ClO2
InChI KeyRZNHSEZOLFEFGB-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear yellow to brown
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.5714 to 1.5734 (20°C, 589 nm)