Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
CAS: 140-88-5 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR117980250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl acrylate
Name NoteStabilized
CAS140-88-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Toxic if inhaled