Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dithiooxamide, 98%, Thermo Scientific Chemicals
CAS: 79-40-3 | C2H4N2S2 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ACR117551000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS4065-45-6
IUPAC Name5-benzoyl-4-hydroxy-2-methoxybenzene-1-sulfonic acid
Molecular FormulaC14H12O6S
InChI KeyCXVGEDCSTKKODG-UHFFFAOYSA-N
SMILESCOC1=CC(O)=C(C=C1S(O)(=O)=O)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥83.0% (dry wt. basis)
Appearance (Color)White to pale yellow
Water Content (Karl Fischer Titration)≤12.75%