Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR116420250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS64628-74-6
IUPAC Name3-chloro-4-[(trifluoromethyl)sulfanyl]aniline
Molecular FormulaC7H5ClF3NS
InChI KeyBLOPJEJSRKMVSU-UHFFFAOYSA-N
SMILESNC1=CC=C(SC(F)(F)F)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder or lumps
Appearance (Color)Pale brown to brown
Melting Point (clear melt)39.0-45.0?C