N,N'-Diisopropylcarbodiimide, 99%, Thermo Scientific Chemicals
N,N'-Diisopropylcarbodiimide, 99%, Thermo Scientific Chemicals
N,N'-Diisopropylcarbodiimide, 99%, Thermo Scientific Chemicals
N,N'-Diisopropylcarbodiimide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N'-Diisopropylcarbodiimide, 99%, Thermo Scientific Chemicals
CAS: 693-13-0 | C7H14N2 | 126.2 g/mol
Have Questions?
Catalog NumberdimQuantityPackaging
ACR115210250
Catalog number ACR115210250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS1898-66-4
IUPAC Name1,1-diphenyl-2-(2,4,6-trinitrophenyl)hydrazine
Molecular FormulaC18H13N5O6
InChI KeyWCBPJVKVIMMEQC-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(=C(NN(C2=CC=CC=C2)C2=CC=CC=C2)C(=C1)[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green to black
Assay (unspecified)≥90.0% ( Elemental analysis or HPLC)
FormPowder
Coupling reagent
RUO – Research Use Only