N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number ACR114425000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16940-66-2
IUPAC Namesodium boranuide
Molecular FormulaBH4Na
InChI KeyYOQDYZUWIQVZSF-UHFFFAOYSA-N
SMILES[BH4-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Lightness91.4 to 100 (lightness scale)
Appearance (Form)Fine powder
Iron (Fe)=<5 ppm
Silicate (as SiO2)0.0021 to 0.30 %
Appearance (Color)White