N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR114421000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS119-61-9
IUPAC Namediphenylmethanone
Molecular FormulaC13H10O
InChI KeyRWCCWEUUXYIKHB-UHFFFAOYSA-N
SMILESO=C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
GC>=99.0 %
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
Melting point47°C to 49°C