1,1-Dichlorodimethyl ether, 97%, Thermo Scientific Chemicals
1,1-Dichlorodimethyl ether, 97%, Thermo Scientific Chemicals
1,1-Dichlorodimethyl ether, 97%, Thermo Scientific Chemicals
1,1-Dichlorodimethyl ether, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,1-Dichlorodimethyl ether, 97%, Thermo Scientific Chemicals
CAS: 4885-02-3 | C2H4Cl2O | 114.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR113471000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 1-Dichlorodimethyl ether
CAS4885-02-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0