1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Diazabicyclo[2.2.2]octane, 97%, Thermo Scientific Chemicals
CAS: 280-57-9 | C6H12N2 | 112.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR11247025025 gGlass Bottle
Catalog number ACR112470250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS589-91-3
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4565-1.4605 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%