1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ACR111175000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS23612-57-9
IUPAC Name(2-aminopyridin-3-yl)methanol
Molecular FormulaC6H8N2O
InChI KeyFEIACFYXEWBKHU-UHFFFAOYSA-N
SMILESNC1=NC=CC=C1CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%