trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
CAS: 14371-10-9 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
500 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number ACR110350500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5071-96-5
IUPAC Name1-(3-methoxyphenyl)methanamine
Molecular FormulaC8H11NO
InChI KeyGRRIMVWABNHKBX-UHFFFAOYSA-N
SMILESCOC1=CC=CC(CN)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5455-1.5495 @ 20°C
FormLiquid
Appearance (Color)Clear colorless to yellow
Identification (FTIR)Conforms
Assay (GC)≥96.0%