1,8-Cineole, 99%, Thermo Scientific Chemicals
1,8-Cineole, 99%, Thermo Scientific Chemicals
1,8-Cineole, 99%, Thermo Scientific Chemicals
1,8-Cineole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,8-Cineole, 99%, Thermo Scientific Chemicals
CAS: 470-82-6 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR110345000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7646-85-7
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration Complexometric97.5 to 102.5 %
Cadmium (Cd)=<10 ppm
Copper (Cu)=<10 ppm