Biuret, 97%, extra pure
Biuret, 97%, extra pure
Biuret, 97%, extra pure
Biuret, 97%, extra pure
Thermo Scientific Chemicals
Biuret, 97%, extra pure
CAS: 108-19-0 | C2H5N3O2 | 103.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Catalog number ACR106520250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS4328-13-6
IUPAC Nametetrahexylazanium bromide
Molecular FormulaC24H52BrN
InChI KeySYZCZDCAEVUSPM-UHFFFAOYSA-M
SMILES[Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormPowder or crystalline powder or flakes
Assay (Titration ex Bromide)≥97.5 to ≤102.5%