Bibenzyl, 99%, Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR106231000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBibenzyl
Name Note99%
CAS103-29-7
CAS Min %98.5
CAS Max %100.0