Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
5 mg
100 mg
Packaging:
Glass bottle
Catalog number ACR105550250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBenzo[ghi]perylene
Name Note98+%
CAS191-24-2
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning