p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
CAS: 104-94-9 | C7H9NO | 123.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1048300505 gGlass bottle
Catalog number ACR104830050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialp-Anisidine
Name Note99%
CAS104-94-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
May cause cancer.
Fatal if inhaled.
Fatal in contact with skin.
Fatal if swallowed.
Very toxic to aquatic life.