D(-)-Amygdalin hydrate, 95%
D(-)-Amygdalin hydrate, 95%
D(-)-Amygdalin hydrate, 95%
D(-)-Amygdalin hydrate, 95%
Thermo Scientific Chemicals
D(-)-Amygdalin hydrate, 95%
CAS: 29883-15-6 | C20H27NO11 | 457.43 g/mol
Have Questions?
Catalog NumberQuantity
ACR1047300505 g
Catalog number ACR104730050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS57-87-4
IUPAC Name(1R,3aR,7S,9aR,9bS,11aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Molecular FormulaC28H44O
InChI KeyDNVPQKQSNYMLRS-APGDWVJJSA-N
SMILESCC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2C3=CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
HPLC>=97.5 %
Appearance (Form)Crystalline powder or needles
Specific optical rotation-105° to -135° (20°C, 589 nm) (c=1, CHCl3)