Nunc™ Seals, Tapes, and Foils
Nunc™ Seals, Tapes, and Foils
Actual product may vary
Nunc™ Seals, Tapes, and Foils
Nunc™ Seals, Tapes, and Foils
Nunc™ Seals, Tapes, and Foils
Thermo Fisher Scientific
Nunc™ Seals, Tapes, and Foils
Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialFor Use With (Application)Certified RNase/DNase-free
AB-0580PVC-coated PolypropyleneCrystallography and genotypingNo
232698AluminumPCR, storage in 96-well platesYes
Catalog number AB-0580
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Material:
PVC-coated Polypropylene
For Use With (Application):
Crystallography and genotyping
Certified RNase/DNase-free:
No
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
Specifications
ColorClear
Solvent Or Alcohol ResistantLife Science Reagent Resistant
DescriptionAdhesive Plate Seals
MaterialPVC-coated Polypropylene
SterilityNon-sterile
AutoclavableAutoclavable
No. of Wells96
No. per Case100
No. per Pack100
For Use With (Application)Crystallography and genotyping
Product TypeAdhesive Seal
Product LineAbgene
AdhesiveAcrylic
Certified RNase/DNase-freeNo
PierceableYes
Temperature Range (Metric)-40°C to +80°C
Well ShapeAll well shapes
Plate CompatibilityAll Microplates
Incubation ConditionsGeneral
PeelableYes
Unit Size
Contents & Storage
General; For long or short term storage