Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
Actual product may vary
Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
Thermo Fisher Scientific
Abgene™ 96-well Sealing Mats for Sample Processing & Storage DeepWell™ and MicroWell Microplates
Round or square CapMats, Sealing Mats, MatCaps for sealing 96-well storage plates, available in autoclavable and piercable formats.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature Range (Metric)Well ShapeFit for Purpose
AB-0566-20°C to +55°CRoundPCR Ready
AB-0674-20°C to +130°CRoundPCR Ready
Catalog number AB-0566
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Temperature Range (Metric):
-20°C to +55°C
Well Shape:
Round
Fit for Purpose:
PCR Ready
Round or square CapMats, Sealing Mats, MatCaps for sealing 96-well storage plates, available in autoclavable and piercable formats.
Thermo Scientific 96-well Sealing Mats are designed for 96-well storage plates and are available in either piercable or autoclavable formats. The round or square capped mats are made of thermoplastic elastomer and are suitable for long-term storage.
Highlights
  • Certified RNAse, DNAse, PCR Inhibitor, and human DNA free
  • Round or square capped mats for sealing 96-well storage plates
  • Alphanumeric grid-referencing
  • Molded from a proprietary thermoplastic elastomer for a flexible design
  • Suitable for long term storage
  • Available in autoclavable or piercable formats
Pierceable version:
  • Solvent resistant (including DMSO, Ethanol, and Isopropanol)
  • Suitable for Gamma Irradiation
  • Requires 6N for piercing
  • Temperature Range: −20°C to +55°C
Autoclavable version:
  • Suitable for Gamma Irradiation
  • Temperature Range: −20°C to +130°C
Hazardous: No
Storage Condition: Ambient
Specifications
ColorNatural
Solvent Or Alcohol ResistantAlcohol Resistant, Solvent Resistant
DescriptionCapMat for 96-well Polypropylene Sample Processing & Storage Plates
Certifications/ComplianceCertified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
MaterialTPE
SterilityCleanroom
AutoclavableNot Autoclavable
No. of Wells96
No. per Case50
No. per Pack10
Plug Diameter7mm
Temperature Range (English)-4°F to 131°F
For Use With (Application)Designed for sealing 96-well storage plate and suitable for long term storage
Format96-well
Product Type96-well capmat, sealing mat
Product LineAbgene
AdhesiveNone
Certified RNase/DNase-freeYes
PierceableYes
Temperature Range (Metric)-20°C to +55°C
Well ShapeRound
Plate CompatibilityAbgene 96 Round Well Plate
Fit for PurposePCR Ready
Unit Size