ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
Actual product may vary
ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
ClipTip™ Filtered 384-Format Pipette Tips
Thermo Fisher Scientific
ClipTip™ Filtered 384-Format Pipette Tips
Thermo Scientific™ ClipTip™ 384-format pipette tips are designed for 384-format microplate applications, in conjunction with the E1-ClipTip electronic pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip Style
9442010330 μLStandard 384
9442005312.5 μLStandard 384
94420153125 μLStandard 384
Catalog number 94420103
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
30 μL
Tip Style:
Standard 384
Thermo Scientific™ ClipTip™ 384-format pipette tips are designed for 384-format microplate applications, in conjunction with the E1-ClipTip electronic pipettes. Featuring an innovativesnap and seal tip attachment mechanism, the pipette tips ensure a light force with a secure tip attachment, enabling greater efficiency and confidence in your work.
Key Features:
  • All ClipTip 384-format tips feature a ‘Snap and Seal’ tip attachment mechanism that ensures light force and secure tip attachment
  • Each tip has incorporated small protrusions that snap on the tip fitting flange of the pipette
  • Ideal for 384-format microplate applications
  • Filtered tips are ideal for sensitive assays, such as PCR, and with critical samples to prevent cross-contaminations
Proven quality
  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Versatile Tip Range
  • 96-format volumes: 12.5, 20, 50, 200, 300, 1000 and 1250 μL
  • 384-format volumes: 12.5, 30 and 125μL
  • Extended lenght tips available for volumes 12.5 and 300 μL allowing for better reach with specific labware consumables
Immediate traceability
  • Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color code of the F1-ClipTip and E1-ClipTip pipette with the corresponding ClipTip pipette tip rack insert
Note:
Availability may vary by country.
Specifications
DescriptionClipTip 384 30, Filtered, Sterile, Rack
Volume (Metric)30 μL
FilteredFiltered
SterilitySterile
AutoclavableNot Autoclavable
Length (English)1.7 in.
Length (Metric)4.4cm
FormatRack
Tip StyleStandard 384
Packaging10 x 384 tips/rack
Color CodeViolet
For Use With (Equipment)Thermo Scientific E1-ClipTip, E1-ClipTip Electronic
Product LineClipTip
Fit for PurposeSterile & PCR Ready
Unit Size3840